Diethyl-bis(hydroxymethyl)malonate 96%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2,2-bis(hydroxymethyl)malonate

A397444-100G A397444-10G A397444-25G A397444-500G A397444-5G
77264-484EA 65.92 USD
77264-484 77264-486 77264-488 77264-490 77264-492
Diethyl-bis(hydroxymethyl)malonate 96%
Diethyl-bis(hydroxymethyl)malonate

Precaución: for Research Use Only

Formula: C₉H₁₆O₆
MW: 220.221 g/mol
MDL Number: MFCD00009130
CAS Number: 20605-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR