2,4,6-Trimethoxybenzyl alcohol 95%
Supplier: Ambeed
|
Warning
|
A396784-250MG
A396784-25G
A396784-5G
77252-084EA
25.75
USD
77252-084
77252-086
77572-220
2,4,6-Trimethoxybenzyl alcohol 95%
2,4,6-Trimethoxybenzyl alcohol
Precaución: for Research Use Only
Formula:
C₁₀H₁₄O₄ MW: 198.22 g/mol |
MDL Number:
MFCD00060356 CAS Number: 61040-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...