(S)-tert-Butyl (3-amino-2-hydroxypropyl)carbamate 95%

Supplier: Ambeed
Warning

A396452-100MG A396452-1G A396452-250MG A396452-50MG
77240-570EA 195.7 USD
77240-570 77240-572 77240-574 77240-576
(S)-tert-Butyl (3-amino-2-hydroxypropyl)carbamate 95%
(S)-tert-Butyl (3-amino-2-hydroxypropyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₃
MW: 190.24 g/mol
MDL Number: MFCD30153192
CAS Number: 853944-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR