4-Aminoantipyrine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

A396271-100MG A396271-25G A396271-1MG A396271-500G
77291-118EA 5.15 USD
77291-118 77616-710 77748-796 77748-797
4-Aminoantipyrine 98%
4-Aminoantipyrine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
MDL Number: MFCD00003145
CAS Number: 83-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR