2-Bromo-5-formylbenzoic acid 97%

Supplier: Ambeed
Warning

A395948-1G A395948-250MG
77264-472EA 130 USD
77264-472 77264-474
2-Bromo-5-formylbenzoic acid 97%
2-Bromo-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
MDL Number: MFCD26855793
CAS Number: 1289007-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR