2,2'-Dithiobis(5-nitropyridine) 96%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(5-nitropyridin-2-yl)disulphane

A395464-100MG A395464-1G A395464-250MG A395464-5G
77252-078EA 17.51 USD
77252-078 77252-080 77252-082 77572-212
2,2'-Dithiobis(5-nitropyridine) 96%
2,2'-Dithiobis(5-nitropyridine)

Precaución: for Research Use Only

Formula: C₁₀H₆N₄O₄S₂
MW: 310.31 g/mol
MDL Number: MFCD00006453
CAS Number: 2127-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR