1-Methyl-1H-indol-2-ol 96%

Supplier: Ambeed
Danger

A395430-250MG A395430-1G
77240-568EA 27.81 USD
77240-568 77746-793
1-Methyl-1H-indol-2-ol 96%
1-Methyl-1H-indol-2-ol

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD01935997
CAS Number: 90563-58-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR