4-Amino-3-methylbenzotrifluoride 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-4-(trifluoromethyl)aniline

A395309-1G A395309-5G A395309-10G A395309-250MG
77278-698EA 53.56 USD
77278-698 77278-700 77572-210 77748-794
4-Amino-3-methylbenzotrifluoride 98%
4-Amino-3-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₈F₃N
MW: 175.15 g/mol
MDL Number: MFCD03701068
CAS Number: 67169-22-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR