4-(Piperidin-4-yl)benzoic acid hydrochloride 98%

Supplier: Ambeed
Warning

A395249-1G A395249-250MG A395249-5G
77291-106EA 64.89 USD
77291-106 77291-108 77291-110
4-(Piperidin-4-yl)benzoic acid hydrochloride 98%
4-(Piperidin-4-yl)benzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂·ClH
MW: 241.714 g/mol
MDL Number: MFCD08460726
CAS Number: 149353-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR