2,4-Dimethoxybenzenesulfonamide 96%

Supplier: Ambeed
Warning

A395156-1G A395156-5G
77252-074EA 25.01 USD
77252-074 77252-076
2,4-Dimethoxybenzenesulfonamide 96%
2,4-Dimethoxybenzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₁NO₄S
MDL Number: MFCD06147001
CAS Number: 51770-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR