Dichlorobis(mu-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetrachlorobis(2,7-dimethyl-2,6-octadienylene)diruthenium, Dichlorobis(μ-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV), Dichlorodi-μ-chlorobis[(1,2,3,6,7,8-η-2,7-dimethyl-2,6-octadiene-1,8-diyl]diruthenium(IV)
A395064-10G
A395064-1G
A395064-250MG
A395064-5G
77252-066EA
721
USD
77252-066
77252-068
77252-070
77252-072
Dichlorobis(mu-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV) 98%
Dichlorobis(mu-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV)
Precaución: for Research Use Only
Formula:
C₂₀H₃₂Cl₄Ru₂ |
MDL Number:
MFCD08459355 CAS Number: 34801-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...