2-Chloroacrylic acid ≥98% stabilized
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Chloroacrylicacid
Stabilization:
0.3-0.5% Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A395021-5G
A395021-25G
77264-462EA
57
USD
77264-462
77551-182
2-Chloroacrylic acid ≥98% stabilized
2-Chloroacrylic acid
Precaución: for Research Use Only
Formula:
C₃H₃ClO₂ MW: 106.51 g/mol |
MDL Number:
MFCD00014336 CAS Number: 598-79-8 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...