3-Mercaptobenzoic acid 97%

Supplier: Ambeed
Danger

A394833-10G A394833-1G A394833-250MG A394833-5G
77278-684EA 206 USD
77278-684 77278-686 77278-688 77278-690
3-Mercaptobenzoic acid 97%
3-Mercaptobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₂S
MW: 154.1863 g/mol
MDL Number: MFCD00238636
CAS Number: 4869-59-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR