1-(3-Amino-5-fluoro-2-methylphenyl)ethanone 97%

Supplier: Ambeed
Warning

A394700-100MG A394700-250MG
77264-458EA 70.04 USD
77264-458 77264-460
1-(3-Amino-5-fluoro-2-methylphenyl)ethanone 97%
1-(3-Amino-5-fluoro-2-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₁₀FNO
MW: 167.18 g/mol
MDL Number: MFCD30189224
CAS Number: 2089292-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR