2-Phenyl-2-oxazoline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Phenyl(2-oxazoline), (4,5-Dihydro-2-oxazolyl)benzene, 4,5-Dihydro-2-phenyloxazole
A394558-100G
A394558-25G
A394558-5G
77291-100EA
175.1
USD
77291-100
77291-102
77291-104
2-Phenyl-2-oxazoline 98%
2-Phenyl-2-oxazoline
Precaución: for Research Use Only
Formula:
C₉H₉NO MW: 147.1739 g/mol |
MDL Number:
MFCD00075541 CAS Number: 7127-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...