Mesitylenesulfonic acid dihydrate 98%

Supplier: Ambeed
Danger

A394439-25G A394439-5G
77240-554EA 39 USD
77240-554 77240-556
Mesitylenesulfonic acid dihydrate 98%
Mesitylenesulfonic acid dihydrate

Precaución: for Research Use Only

Formula: C₉H₁₂O₃S·2H₂O
MDL Number: MFCD00149535
CAS Number: 835617-36-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR