1,1,2,2-Tetrakis(4-(1H-imidazol-1-yl)phenyl)ethene 97%

Supplier: Ambeed
Warning

A394434-1G A394434-250MG A394434-5G
77264-450EA 48.41 USD
77264-450 77264-452 77264-454
1,1,2,2-Tetrakis(4-(1H-imidazol-1-yl)phenyl)ethene 97%
1,1,2,2-Tetrakis(4-(1H-imidazol-1-yl)phenyl)ethene

Precaución: for Research Use Only

Formula: C₃₈H₂₈N₈
MW: 596.68 g/mol
CAS Number: 1477527-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR