4,4'-Difluoro-2,2'-bipyridine 97%
Supplier: Ambeed
|
Warning
|
A394119-100MG
A394119-1G
A394119-250MG
77278-676EA
124
USD
77278-676
77278-678
77278-680
4,4'-Difluoro-2,2'-bipyridine 97%
4,4'-Difluoro-2,2'-bipyridine
Precaución: for Research Use Only
Formula:
C₁₀H₆F₂N₂ |
MDL Number:
MFCD11042493 CAS Number: 1189458-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...