2-(4-Fluorophenyl)-4-methylthiazole-5-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A394076-1G
A394076-250MG
77240-536EA
56.65
USD
77240-536
77240-538
2-(4-Fluorophenyl)-4-methylthiazole-5-carboxylic acid 95%
2-(4-Fluorophenyl)-4-methylthiazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₈FNO₂S MW: 237.25 g/mol |
MDL Number:
MFCD06857923 CAS Number: 144060-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...