2-Bromo-4,6-difluoroaniline 96%

Supplier: Ambeed
Danger

A393713-25G A393713-5G
77291-096EA 18 USD
77291-096 77291-098
2-Bromo-4,6-difluoroaniline 96%
2-Bromo-4,6-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrF₂N
MDL Number: MFCD00009639
CAS Number: 444-14-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR