ICA 069673 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-Chloropyrimidin-5-yl)-3,4-difluorobenzamide

A393175-10MG A393175-1MG A393175-25MG A393175-50MG A393175-5MG
77278-654EA 125.66 USD
77278-654 77278-656 77278-658 77278-660 77278-662
ICA 069673 98%
ICA 069673

Precaución: for Research Use Only

Formula: C₁₁H₆ClF₂N₃O
MW: 269.63 g/mol
MDL Number: MFCD20926351
CAS Number: 582323-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR