(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine 98%

Supplier: Ambeed
Warning

Synonyms: (11aS)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine, (S)-SIPHOS-PE

A393102-50MG A393102-250MG
77252-050EA 100.94 USD
77252-050 77616-690
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine 98%
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine

Precaución: for Research Use Only

Formula: C₃₃H₃₂NO₂P
MW: 505.58 g/mol
MDL Number: MFCD08459342
CAS Number: 500997-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR