2-Amino-1-(pyrrolidin-1-yl)ethanone 98%

Supplier: Ambeed
Warning

A392423-100MG A392423-1G A392423-250MG
77278-628EA 33 USD
77278-628 77278-630 77278-632
2-Amino-1-(pyrrolidin-1-yl)ethanone 98%
2-Amino-1-(pyrrolidin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MDL Number: MFCD08060127
CAS Number: 24152-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR