4-Bromo-1,2-dimethylimidazole 96%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-1,2-dimethyl-1H-imidazole

A392247-100MG A392247-1G A392247-250MG A392247-5G
77291-074EA 27.81 USD
77291-074 77291-076 77291-078 77291-080
4-Bromo-1,2-dimethylimidazole 96%
4-Bromo-1,2-dimethylimidazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂
MW: 175.03 g/mol
MDL Number: MFCD06659901
CAS Number: 850429-59-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR