Bromo-PEG3-bromide 96%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-(2-(2-(2-bromoethoxy)ethoxy)ethoxy)ethane

A392199-100MG A392199-1G A392199-250MG A392199-25G A392199-5G A392199-100G
77264-426EA 5 USD
77264-426 77264-428 77264-430 77264-432 77264-434 77748-787
Bromo-PEG3-bromide 96%
Bromo-PEG3-bromide

Precaución: for Research Use Only

Formula: C₈H₁₆Br₂O₃
MDL Number: MFCD18205853
CAS Number: 31255-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR