(S)-2-Aminopropanoic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A392150-1G
A392150-250MG
A392150-5G
77264-420EA
7
USD
77264-420
77264-422
77264-424
(S)-2-Aminopropanoic acid hydrochloride 97%
(S)-2-Aminopropanoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₈ClNO₂ |
MDL Number:
MFCD03791086 CAS Number: 6003-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...