HPOB 97%

Supplier: Ambeed
Danger

Synonyms: N-Hydroxy-4-(2-((2-hydroxyethyl)(phenyl)amino)-2-oxoethyl)benzamide

A392083-10MG A392083-1MG A392083-25MG A392083-5MG
77264-412EA 146.26 USD
77264-412 77264-414 77264-416 77264-418
HPOB 97%
HPOB

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₄
MW: 314.34 g/mol
MDL Number: MFCD27952937
CAS Number: 1429651-50-2
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR