2-Chloro-4-cyanobenzenesulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-4-cyanobenzene-1-sulphonyl chloride

A391492-100MG A391492-1G A391492-250MG A391492-5G
77264-402EA 7 USD
77264-402 77264-404 77264-406 77264-408
2-Chloro-4-cyanobenzenesulphonyl chloride 98%
2-Chloro-4-cyanobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₂S
MW: 236.08
MDL Number: MFCD00177089
CAS Number: 254749-11-6
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR