1-(4-Fluoro-3-methoxyphenyl)thiourea 97%

Supplier: Ambeed
Danger

A391450-1G A391450-250MG
77240-510EA 73.13 USD
77240-510 77240-512
1-(4-Fluoro-3-methoxyphenyl)thiourea 97%
1-(4-Fluoro-3-methoxyphenyl)thiourea

Precaución: for Research Use Only

Formula: C₈H₉FN₂OS
MW: 200.23 g/mol
MDL Number: MFCD18379875
CAS Number: 1237759-54-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR