Diethyl 2,4-Pyridinedicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl pyridine-2,4-dicarboxylate, 2,4-Pyridinedicarboxylic acid Diethyl ester

A391300-1G A391300-25G A391300-5G
77252-030EA 120.51 USD
77252-030 77252-032 77252-034
Diethyl 2,4-Pyridinedicarboxylate 97%
Diethyl 2,4-Pyridinedicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
MDL Number: MFCD00152168
CAS Number: 41438-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR