(R)-3-Amino-3-phenylpropionic acid 97%

Supplier: Ambeed
Warning

Synonyms: H-D-β-Phenyl-Ala-OH, 3-Amino-3-phenylpropionic-acid, (R)-3-Amino-3-phenylpropanoic acid, (R)-b-Amino-benzenepropanoic acid

A390776-10G A390776-1G A390776-250MG A390776-25G A390776-5G
77252-020EA 221.45 USD
77252-020 77252-022 77252-024 77252-026 77252-028
(R)-3-Amino-3-phenylpropionic acid 97%
(R)-3-Amino-3-phenylpropionic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
MDL Number: MFCD01076237
CAS Number: 13921-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR