3-Cyano-3-phenylpropanoic acid 97%

Supplier: Ambeed
Warning

A390691-250MG A390691-1G A390691-5G
77278-606EA 33 USD
77278-606 77594-192 77748-786
3-Cyano-3-phenylpropanoic acid 97%
3-Cyano-3-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD03032627
CAS Number: 14387-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR