1-Benzyloxy-2-bromobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-benzyloxybenzene, 1-(Benzyloxy)-2-bromobenzene

A390688-100G A390688-5G
77264-390EA 123.6 USD
77264-390 77264-392
1-Benzyloxy-2-bromobenzene 98%
1-Benzyloxy-2-bromobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
MDL Number: MFCD03427006
CAS Number: 31575-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR