Glycerol monostearate 98%

Supplier: Ambeed
Warning

A390484-100G A390484-25G
77240-506EA 11 USD
77240-506 77240-508
Glycerol monostearate 98%
Glycerol monostearate

Precaución: for Research Use Only

Formula: C₂₁H₄₄O₅
MDL Number: MFCD00036186
CAS Number: 31566-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR