(R)-2,2'-Bis(dicyclohexylphosphino)-1,1'-binaphthalene 98%

Supplier: Ambeed
Warning

A390449-100MG A390449-1G A390449-250MG A390449-50MG
77264-384EA 166.86 USD
77264-384 77264-386 77264-388 77616-680
(R)-2,2'-Bis(dicyclohexylphosphino)-1,1'-binaphthalene 98%
(R)-2,2'-Bis(dicyclohexylphosphino)-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₄₄H₅₆P₂
MW: 646.86 g/mol
MDL Number: MFCD01630804
CAS Number: 139139-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR