1-Cyclopentylethanone 98%

Supplier: Ambeed
Danger

A390410-1G A390410-25G A390410-5G
77240-500EA 7 USD
77240-500 77240-502 77240-504
1-Cyclopentylethanone 98%
1-Cyclopentylethanone

Precaución: for Research Use Only

Formula: C₇H₁₂O
MDL Number: MFCD00060799
CAS Number: 6004-60-0
UN: 1224
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR