(2,3-Dichloro-6-nitrophenyl)methanamine 95%

Supplier: Ambeed
Danger

A390375-100MG A390375-1G A390375-250MG
77252-010EA 47.38 USD
77252-010 77252-012 77252-014
(2,3-Dichloro-6-nitrophenyl)methanamine 95%
(2,3-Dichloro-6-nitrophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MW: 221.04 g/mol
MDL Number: MFCD01075037
CAS Number: 70380-49-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR