(+)-6,6'-{[(1R,3R)-1,3-Dimethyl-1,3-propanediyl]bis(oxy)}bis[4,8-bis(t-butyl)-2,10-dimethoxy-bibenzo[d,f][1,3,2]dioxaphosphepin] 98%

Supplier: Ambeed
Warning

A390314-250MG A390314-1G
77252-008EA 137 USD
77252-008 77551-154
(+)-6,6'-{[(1R,3R)-1,3-Dimethyl-1,3-propanediyl]bis(oxy)}bis[4,8-bis(t-butyl)-2,10-dimethoxy-bibenzo[d,f][1,3,2]dioxaphosphepin] 98%
(+)-6,6'-{[(1R,3R)-1,3-Dimethyl-1,3-propanediyl]bis(oxy)}bis[4,8-bis(t-butyl)-2,10-dimethoxy-bibenzo[d,f][1,3,2]dioxaphosphepin]

Precaución: for Research Use Only

Formula: C₄₉H₆₆O₁₀P₂
MDL Number: MFCD06798296
CAS Number: 149646-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR