(2-Formyl-4-nitrophenoxy)acetic acid 97%

Supplier: Ambeed
Warning

A390155-1G A390155-5G
77240-496EA 22 USD
77240-496 77240-498
(2-Formyl-4-nitrophenoxy)acetic acid 97%
(2-Formyl-4-nitrophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₆
MDL Number: MFCD00024502
CAS Number: 6965-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR