2-[(Dimethylamino)methyl]benzonitrile 97%

Supplier: Ambeed
Warning

A389511-1G A389511-5G
77252-002EA 53 USD
77252-002 77252-004
2-[(Dimethylamino)methyl]benzonitrile 97%
2-[(Dimethylamino)methyl]benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂
MDL Number: MFCD07324818
CAS Number: 53369-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR