4-Chloro-3-nitrobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Nitro-4-chlorobenzenesulphonamide

A389498-100G A389498-10G A389498-1G A389498-500G A389498-5G
77291-054EA 32.96 USD
77291-054 77291-056 77291-058 77291-060 77291-062
4-Chloro-3-nitrobenzenesulfonamide 98%
4-Chloro-3-nitrobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₄S
MW: 236.633 g/mol
MDL Number: MFCD00035783
CAS Number: 97-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR