3-Fluoro-2-iodoanisole 95%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoro-6-methoxyiodobenzene, 3-Fluoro-2-iodophenyl methyl ether, 1-Fluoro-2-iodo-3-methoxybenzene

A389275-1G A389275-250MG A389275-5G
77278-594EA 49.44 USD
77278-594 77278-596 77278-598
3-Fluoro-2-iodoanisole 95%
3-Fluoro-2-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆FIO
MW: 252.025 g/mol
MDL Number: MFCD03788539
CAS Number: 7079-54-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR