Cyclopropyl(4-(trifluoromethyl)phenyl)methanamine 97%

Supplier: Ambeed
Warning

A389130-100MG A389130-1G A389130-250MG
77251-990EA 101.97 USD
77251-990 77251-992 77251-994
Cyclopropyl(4-(trifluoromethyl)phenyl)methanamine 97%
Cyclopropyl(4-(trifluoromethyl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₃N
MW: 215.21 g/mol
MDL Number: MFCD11848970
CAS Number: 581812-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR