1-(4-((Trifluoromethyl)thio)phenyl)ethanamine 97%

Supplier: Ambeed
Warning

A388761-100MG A388761-250MG
77251-986EA 112.27 USD
77251-986 77251-988
1-(4-((Trifluoromethyl)thio)phenyl)ethanamine 97%
1-(4-((Trifluoromethyl)thio)phenyl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₀F₃NS
MW: 221.24 g/mol
MDL Number: MFCD18703057
CAS Number: 871519-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR