4'-Methyl-[2,2'-bipyridine]-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A388272-100MG A388272-250MG A388272-1G
77240-476EA 54.59 USD
77240-476 77240-478 77746-779
4'-Methyl-[2,2'-bipyridine]-4-carbaldehyde 97%
4'-Methyl-[2,2'-bipyridine]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O
MW: 198.22 g/mol
MDL Number: MFCD11112058
CAS Number: 104704-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR