6-Methoxy-o-cresol 97%
Supplier: Ambeed
|
Warning
|
A388180-100G
A388180-1G
A388180-250MG
A388180-25G
A388180-5G
77264-338EA
304.01
USD
77264-338
77264-340
77264-342
77264-344
77264-346
6-Methoxy-o-cresol 97%
6-Methoxy-o-cresol
Precaución: for Research Use Only
Formula:
C₈H₁₀O₂ |
MDL Number:
MFCD00043771 CAS Number: 2896-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...