1-(3,5-Dibromopyridin-4-yl)ethanone 95%

Supplier: Ambeed
Warning

A387937-1G A387937-250MG A387937-5G
77264-324EA 309 USD
77264-324 77264-326 77594-180
1-(3,5-Dibromopyridin-4-yl)ethanone 95%
1-(3,5-Dibromopyridin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO
MW: 278.92 g/mol
MDL Number: MFCD18910177
CAS Number: 870244-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR