2,6-Difluoroanisole 98%

Supplier: Ambeed
Danger

A387312-1G A387312-25G A387312-5G
77251-980EA 10 USD
77251-980 77251-982 77251-984
2,6-Difluoroanisole 98%
2,6-Difluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆F₂O
MDL Number: MFCD00142846
CAS Number: 437-82-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR