GRA Ex-25 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-((1-(trans-4-(tert-Butyl)cyclohexyl)-3-(4-(trifluoromethoxy)phenyl)ureido)methyl)benzamido)propanoic acid

A387310-10MG A387310-1MG A387310-2MG A387310-5MG
77264-316EA 247.2 USD
77264-316 77264-318 77264-320 77264-322
GRA Ex-25 98%
GRA Ex-25

Precaución: for Research Use Only

Formula: C₂₉H₃₆F₃N₃O₅
MW: 563.60 g/mol
MDL Number: MFCD11977277
CAS Number: 307983-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR