5-Azacytidine 95%

Supplier: Ambeed
Warning

Synonyms: Ladakamycin, 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one

A386837-1G A386837-1MG A386837-250MG A386837-25G A386837-5G A386837-100G
77278-584EA 20.6 USD
77278-584 77278-586 77278-588 77278-590 77278-592 77748-780
5-Azacytidine 95%
5-Azacytidine

Precaución: for Research Use Only

Formula: C₈H₁₂N₄O₅
MW: 244.21 g/mol
MDL Number: MFCD00006539
CAS Number: 320-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR